Assay Details
Target Gene Details
Entrez Gene ID: | 10522 |
Gene Name: | DEAF1, transcription factor |
Gene Aliases: |
MRD24, NUDR, SPN, ZMYND5 |
Location: |
Chr.11:644220-695754 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DEAF1 | NM_001293634.1 | 1 | 257 | NP_001280563.1 |
NM_021008.3 | 1 | 257 | NP_066288.2 | |
AK291383.1 | ||||
BI463261.1 | 1 | 121 | ||
BM696110.1 | 1 | 243 | ||
HY094363.1 | 1 | 257 |
Target Gene Details
Entrez Gene ID: | 283232 |
Gene Name: | transmembrane protein 80 |
Gene Aliases: |
- |
Location: |
Chr.11:695386-705028 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM80 | XM_017017600.1 | 1 | 113 | XP_016873089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625074 | Chr.11:669311 - 702135 on Build GRCh38 | Loss |
![]() ![]() |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832043 | Chr.11:631027 - 758602 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)