Assay Details
Target Gene Details
Entrez Gene ID: | 11274 |
Gene Name: | ubiquitin specific peptidase 18 |
Gene Aliases: |
ISG43, UBP43 |
Location: |
Chr.22:18149959-18177397 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP18 | NM_017414.3 | 1 | 102 | NP_059110.2 |
XM_006724074.2 | 1 | 134 | XP_006724137.1 | |
AI222587.1 | 2 | 316 | ||
BI752849.1 | ||||
BU584005.1 | 1 | 139 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516997 | Chr.22:18143870 - 19039100 on Build GRCh38 | Gain+Loss | FAM230A DGCR6 GGT3P TMEM191B LOC102725072 LOC100996432 LOC100996415 LOC100996401 LOC729461 PRODH RIMBP3 DGCR9 DGCR5 LOC642643 PI4KAP1 DGCR10 DGCR2 USP18 |
nsv1055756 | Chr.22:18062621 - 18794021 on Build GRCh38 | Gain | FAM230A TUBA8 LOC100996401 GGT3P LOC729461 TMEM191B RIMBP3 PEX26 LOC642643 PI4KAP1 LOC100996415 USP18 |
nsv1058565 | Chr.22:18060494 - 18159378 on Build GRCh38 | Gain | TUBA8 PEX26 USP18 |
esv2751935 | Chr.22:18149609 - 19042823 on Build GRCh38 | Loss | FAM230A DGCR6 GGT3P TMEM191B LOC102725072 LOC100996432 LOC100996415 LOC100996401 LOC729461 PRODH RIMBP3 DGCR9 DGCR5 LOC642643 PI4KAP1 DGCR10 DGCR2 USP18 |
esv3893430 | Chr.22:18059545 - 18169093 on Build GRCh38 | Gain | TUBA8 PEX26 USP18 |
esv2758824 | Chr.22:18069029 - 19074200 on Build GRCh38 | Gain+Loss | FAM230A DGCR11 TUBA8 DGCR6 GGT3P TMEM191B PEX26 LOC102725072 LOC100996432 LOC100996415 LOC100996401 LOC729461 PRODH RIMBP3 DGCR9 DGCR5 LOC642643 PI4KAP1 DGCR10 DGCR2 USP18 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | GGT3P LOC101929372 LOC100996432 LOC100996415 FLJ41941 MIR3198-1 BCL2L13 PI4KAP1 ATP6V1E1 MIR648 USP18 FAM230A TUBA8 TMEM191B PEX26 LOC102725072 BID SLC25A18 CECR2 LOC100996401 LOC729461 RIMBP3 MICAL3 LINC00528 LOC642643 |
nsv471594 | Chr.22:18076012 - 18882129 on Build GRCh38 | Gain+Loss | FAM230A TUBA8 GGT3P TMEM191B PEX26 LOC102725072 LOC100996432 LOC100996415 LOC100996401 LOC729461 RIMBP3 LOC642643 PI4KAP1 USP18 |
nsv469560 | Chr.22:18076011 - 18213970 on Build GRCh38 | Gain | TUBA8 PEX26 LOC100996415 USP18 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |