Assay Details
Target Gene Details
Entrez Gene ID: | 2638 |
Gene Name: | GC, vitamin D binding protein |
Gene Aliases: |
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP |
Location: |
Chr.4:71741693-71805520 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GC | NM_000583.3 | 5 | 905 | NP_000574.2 |
NM_001204306.1 | 6 | 776 | NP_001191235.1 | |
NM_001204307.1 | 6 | 713 | NP_001191236.1 | |
XM_006714177.2 | 5 | 768 | XP_006714240.1 | |
AK223458.1 | 6 | 776 | BAD97178.1 | |
AK290827.1 | 5 | 615 | ||
AK298433.1 | 4 | 473 | ||
AK309595.1 | 6 | 713 | ||
AK312971.1 | 5 | 619 | ||
AK315853.1 | 5 | 768 | ||
BC057228.1 | 5 | 623 | AAH57228.1 | |
EU794674.1 | 5 | 714 | ||
X03178.1 | 5 | 714 | CAA26938.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |