Assay Details
Target Gene Details
Entrez Gene ID: | 1954 |
Gene Name: | multiple EGF like domains 8 |
Gene Aliases: |
C19orf49, CRPT2, EGFL4, SBP1 |
Location: |
Chr.19:42325609-42378769 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEGF8 | NM_001271938.1 | NP_001258867.1 | ||
NM_001410.2 | NP_001401.2 | |||
AB011541.2 | BAA32469.2 | |||
AY280362.1 | AAP35084.1 | |||
BC153880.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | CIC LOC101930071 TMEM145 PRR19 ERF RABAC1 ZNF574 ZNF526 POU2F2 LIPE ATP1A3 GRIK5 CNFN CD79A PAFAH1B3 MIR6797 LOC100505622 DEDD2 MIR4323 ARHGEF1 LIPE-AS1 LOC100505585 MEGF8 LOC390937 RPS19 GSK3A MIR8077 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | PAFAH1B3 CIC LOC101930071 TMEM145 DEDD2 PRR19 LIPE-AS1 ERF MEGF8 ZNF526 POU2F2 LIPE GSK3A CEACAM8 MIR8077 CXCL17 CEACAM1 CNFN |
esv3574530 | Chr.19:42343576 - 42359694 on Build GRCh38 | Gain | MIR8077 MEGF8 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | CIC LOC101930071 TMEM145 PRR19 ERF RABAC1 ZNF574 ZNF526 POU2F2 LIPE CEACAM5 LYPD4 CEACAM6 ATP1A3 GRIK5 CNFN CD79A DMRTC2 PAFAH1B3 CEACAM7 MIR6797 CEACAM3 LOC100505622 DEDD2 MIR4323 ARHGEF1 LIPE-AS1 LOC100505585 MEGF8 CEACAM4 LOC390937 RPS19 GSK3A MIR8077 |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | PAFAH1B3 PSG8 CIC PSG1 LOC101930071 PSG10P TMEM145 LOC100289650 PRR19 PSG2 LIPE-AS1 ERF MEGF8 PSG11 PSG7 PSG6 LIPE PSG3 GSK3A CEACAM8 MIR8077 CXCL17 CEACAM1 CNFN |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |