Assay Details
Target Gene Details
Entrez Gene ID: | 10525 |
Gene Name: | hypoxia up-regulated 1 |
Gene Aliases: |
GRP-170, Grp170, HSP12A, ORP-150, ORP150 |
Location: |
Chr.11:119044185-119057246 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYOU1 | NM_001130991.2 | 23 | 2787 | NP_001124463.1 |
NM_006389.4 | 23 | 2920 | NP_006380.1 | |
XM_005271392.3 | 23 | 2781 | XP_005271449.1 | |
XM_005271393.3 | 23 | 2761 | XP_005271450.1 | |
XM_005271394.3 | 23 | 2828 | XP_005271451.1 | |
XM_017017095.1 | 23 | 2816 | XP_016872584.1 | |
XM_017017096.1 | 23 | 2837 | XP_016872585.1 | |
XM_017017097.1 | 23 | 2760 | XP_016872586.1 | |
AK127324.1 | 22 | 2789 | ||
AK222756.1 | 23 | 2813 | BAD96476.1 | |
AK294905.1 | 23 | 2762 | ||
AK297260.1 | ||||
AK299648.1 | 16 | 1711 | ||
AK301514.1 | 21 | 2479 | ||
AK314178.1 | 23 | 2811 | ||
BC004560.2 | AAH04560.1 | |||
BC072436.1 | AAH72436.1 | |||
CR749610.1 | 3 | 252 | CAH18405.1 | |
DQ350134.1 | 23 | 2808 | ABC75106.1 | |
DQ372932.1 | 23 | 2808 | ABD14370.1 | |
U65785.1 | 23 | 2788 | AAC50947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952745 | Chr.11:119041291 - 119047288 on Build GRCh38 | Deletion | HYOU1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |