Assay Details
Target Gene Details
Entrez Gene ID: | 1798 |
Gene Name: | dolichyl-phosphate N-acetylglucosaminephosphotransferase 1 |
Gene Aliases: |
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT |
Location: |
Chr.11:119096503-119102414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPAGT1 | NM_001382.3 | NP_001373.2 | ||
XM_005271422.2 | XP_005271479.1 | |||
XM_011542648.1 | XP_011540950.1 | |||
XM_017017293.1 | XP_016872782.1 | |||
XM_017017294.1 | XP_016872783.1 | |||
XM_017017295.1 | XP_016872784.1 | |||
AK095122.1 | 3 | 753 | ||
AK095718.1 | ||||
AK128572.1 | ||||
AK225994.1 | ||||
AK301973.1 | ||||
AK312783.1 | ||||
BC000325.2 | AAH00325.1 | |||
BT006802.1 | AAP35448.1 | |||
Z82022.1 | CAB04787.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |