Assay Details
Target Gene Details
Entrez Gene ID: | 199223 |
Gene Name: | tetratricopeptide repeat domain 21A |
Gene Aliases: |
IFT139A, STI2 |
Location: |
Chr.3:39107661-39139009 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTC21A | NM_001105513.2 | NP_001098983.2 | ||
NM_145755.2 | NP_665698.2 | |||
XM_005264921.4 | XP_005264978.1 | |||
XM_005264922.4 | XP_005264979.1 | |||
XM_005264923.3 | XP_005264980.1 | |||
XM_005264924.4 | XP_005264981.1 | |||
XM_005264925.4 | XP_005264982.1 | |||
XM_005264926.4 | XP_005264983.1 | |||
XM_006713011.3 | XP_006713074.1 | |||
XM_011533447.2 | XP_011531749.1 | |||
XM_011533449.1 | XP_011531751.1 | |||
XM_017005843.1 | XP_016861332.1 | |||
AJ487015.1 | CAD31647.1 | |||
AK022483.1 | ||||
AK302408.1 | ||||
AK302465.1 | ||||
DB094952.1 | ||||
DC403032.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |