Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMDHD2 | NM_001145815.1 | NP_001139287.1 | ||
NM_015944.3 | 10 | 1377 | NP_057028.2 | |
XM_017023262.1 | 11 | 1298 | XP_016878751.1 | |
XM_017023263.1 | 9 | 1286 | XP_016878752.1 | |
XM_017023264.1 | 10 | 1229 | XP_016878753.1 | |
XM_017023265.1 | 10 | 1196 | XP_016878754.1 | |
XM_017023266.1 | 10 | 1244 | XP_016878755.1 | |
XM_017023267.1 | 9 | 1240 | XP_016878756.1 | |
AB209351.1 | 5 | 5000 | BAD92588.1 | |
AF132948.1 | 11 | 1268 | AAD27723.1 | |
AK296877.1 | ||||
AK300747.1 | ||||
BC018734.1 | 10 | 1313 | AAH18734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PRR35 CEMP1 MIR3177 RAB26 TBC1D24 MSRB1 RGS11 SNORA10 ABCA3 FAM173A HBZ WDR24 CAPN15 SNRNP25 LOC105371038 LUC7L RPL23AP5 PGP NTN3 LOC100134368 DECR2 MIR4717 BRICD5 DNASE1L2 MIR3178 RAB11FIP3 MIR4516 IFT140 ECI1 NTHL1 NPW RNF151 RHBDL1 CLCN7 CCDC154 HBM RHOT2 CACNA1H MAPK8IP3 LOC105371046 MIR662 UBE2I SNORA78 GNPTG PTX4 MPG NOXO1 TPSAB1 LOC101929280 MIR3180-5 TMEM204 HAGHL TPSG1 NME3 DDX11L10 LOC107984876 WDR90 MCRIP2 RAB40C CCDC78 TPSB2 ATP6V0C WFIKKN1 MEIOB CASKIN1 METTL26 CHTF18 NHLRC4 RPS2 ABCA17P RHBDF1 SPSB3 MIR3176 NUBP2 LOC105371049 AMDHD2 LOC100287175 SNHG9 KCTD5 TPSD1 SNORD60 PRSS27 LMF1-AS1 SYNGR3 EME2 LOC729652 POLR3K AXIN1 IGFALS MSLN TMEM8A SOX8 LINC00235 RNPS1 LOC105371045 LMF1 TBL3 HAGH SLC9A3R2 MIR6859-4 NDUFB10 HBA1 PDPK1 METRN LOC105371184 WASIR2 E4F1 MRPL28 C1QTNF8 MRPS34 NARFL C16orf59 TSC2 LINC00254 FAHD1 C16orf91 FLJ42627 HS3ST6 MIR6768 LOC106660606 MIR940 PIGQ LOC105371047 NPRL3 FAM234A SNORA64 STUB1 SSTR5 MIR6767 HBA2 FBXL16 MIR6511B1 PDIA2 NME4 TRAF7 CCNF PRR25 PKD1 SNHG19 TELO2 CRAMP1 LOC107987233 UNKL ERVK13-1 ARHGDIG HBQ1 MIR1225 RPUSD1 BAIAP3 SSTR5-AS1 TSR3 ZNF598 GNG13 HN1L JMJD8 MIR5587 GFER LOC652276 PRSS29P RPL3L MIR3677 MLST8 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | PRSS33 CEMP1 SRRM2 ATP6V0C SRRM2-AS1 PRSS27 FLJ42627 MIR3178 PRSS41 TBC1D24 PDPK1 TCEB2 ERVK13-1 AMDHD2 LOC652276 KCTD5 PRSS21 |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | CEMP1 ATP6V0C FLJ42627 MIR3178 AMDHD2 TBC1D24 PDPK1 LOC652276 ERVK13-1 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | CEMP1 RAB26 SNORD60 TBC1D24 MIR6768 LOC106660606 MIR940 LOC729652 MIR3180-5 ABCA3 MIR6767 RNPS1 MIR6511B1 TRAF7 CCNF ATP6V0C PGP NTN3 PKD1 SNHG19 CASKIN1 MIR4717 BRICD5 DNASE1L2 MIR3178 PDPK1 ABCA17P MIR4516 MIR1225 ECI1 LOC105371049 E4F1 AMDHD2 LOC652276 MIR3677 MLST8 C16orf59 TSC2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |