Assay Details
Target Gene Details
Entrez Gene ID: | 51104 |
Gene Name: | abhydrolase domain containing 17B |
Gene Aliases: |
C9orf77, CGI-67, FAM108B1 |
Location: |
Chr.9:71859461-71911245 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD17B | NM_001025780.2 | 3 | 1175 | NP_001020951.1 |
NM_016014.3 | 3 | 1175 | NP_057098.2 | |
XM_006717134.3 | 3 | 626 | XP_006717197.2 | |
XM_006717135.1 | 3 | 1163 | XP_006717198.1 | |
XM_017014787.1 | 3 | 782 | XP_016870276.1 | |
XM_017014788.1 | 4 | 1567 | XP_016870277.1 | |
XM_017014789.1 | 3 | 713 | XP_016870278.1 | |
XM_017014790.1 | XP_016870279.1 | |||
XM_017014791.1 | XP_016870280.1 | |||
AF151825.1 | 3 | 859 | AAD34062.1 | |
AK293060.1 | 3 | 1175 | ||
BC038390.1 | 3 | 672 | ||
BC044576.1 | 3 | 1123 | AAH44576.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |