Assay Details
Target Gene Details
Entrez Gene ID: | 4839 |
Gene Name: | NOP2 nucleolar protein |
Gene Aliases: |
NOL1, NOP120, NSUN1, p120 |
Location: |
Chr.12:6556870-6568332 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOP2 | NM_001033714.2 | 15 | 1890 | NP_001028886.1 |
NM_001258308.1 | 15 | 1834 | NP_001245237.1 | |
NM_001258309.1 | 16 | 1933 | NP_001245238.1 | |
NM_001258310.1 | 15 | 1822 | NP_001245239.1 | |
NM_006170.3 | 15 | 1822 | NP_006161.2 | |
XM_005253691.1 | 15 | 1867 | XP_005253748.1 | |
AK056208.1 | 15 | 1804 | ||
AK129852.1 | 15 | 1765 | ||
AK307582.1 | 15 | 1816 | ||
BC000656.2 | 15 | 1755 | AAH00656.1 | |
BC065257.1 | 15 | 1832 | ||
BC082985.1 | 15 | 1772 | ||
BC106072.1 | 16 | 1879 | AAI06073.1 | |
BC128183.1 | 15 | 1809 | ||
BC128184.1 | 15 | 1809 | ||
M32110.1 | 14 | 1748 | AAA36398.1 | |
X55504.1 | 14 | 1704 | CAA39119.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |