Assay Details
Target Gene Details
Entrez Gene ID: | 10279 |
Gene Name: | protease, serine 16 |
Gene Aliases: |
TSSP |
Location: |
Chr.6:27247701-27256620 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS16 | NM_005865.3 | 11 | 1431 | NP_005856.1 |
XM_017010161.1 | 10 | 1353 | XP_016865650.1 | |
XM_017010162.1 | 10 | 1251 | XP_016865651.1 | |
XM_017010163.1 | 10 | 1140 | XP_016865652.1 | |
XM_017010164.1 | 9 | 960 | XP_016865653.1 | |
XM_017010165.1 | 5 | 682 | XP_016865654.1 | |
AF052514.1 | 11 | 1428 | AAC33563.1 | |
AK093590.1 | 2 | 1320 | ||
AK097918.1 | 5 | 682 | ||
AK098033.1 | 2 | 607 | ||
AK126160.1 | 2 | 1973 | ||
AK226091.1 | 2 | 451 | ||
AK303106.1 | 6 | 752 | ||
AK314037.1 | 11 | 1431 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |