Assay Details
Target Gene Details
Entrez Gene ID: | 7525 |
Gene Name: | YES proto-oncogene 1, Src family tyrosine kinase |
Gene Aliases: |
HsT441, P61-YES, Yes, c-yes |
Location: |
Chr.18:721592-812326 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YES1 | NM_005433.3 | 2 | 432 | NP_005424.1 |
XM_005258139.3 | 2 | 394 | XP_005258196.1 | |
XM_017025960.1 | 2 | 274 | XP_016881449.1 | |
AK314063.1 | 2 | 376 | ||
BC048960.1 | 2 | 428 | AAH48960.1 | |
M15990.1 | 2 | 418 | AAA35735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576236 | Chr.18:319176 - 794272 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv5810n54 | Chr.18:351004 - 2238246 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576242 | Chr.18:567127 - 841320 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv528169 | Chr.18:551688 - 2240220 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060188 | Chr.18:575087 - 863351 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2761984 | Chr.18:724222 - 855263 on Build GRCh38 | Loss |
![]() |
nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576228 | Chr.18:69836 - 1506323 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)