Assay Details
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | NP_001157032.1 | ||
NM_152764.2 | NP_689977.2 | |||
BC029829.1 | AAH29829.1 | |||
CD300613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss | LINC00254 HS3ST6 FAHD1 MEIOB |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | HAGH FAHD1 MEIOB |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | HAGH FAHD1 MEIOB |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | ERVK13-1 WDR90 LOC105371047 FAHD1 MIR3677 SOX8 FLJ42627 RGS11 PDIA2 MIR6511B1 MIR4717 LOC106660606 HBZ IFT140 ATP6V0C LUC7L TBL3 MIR940 RPUSD1 DNASE1L2 PTX4 NUBP2 HBA1 LOC105371049 C16orf59 METRN RAB40C HBM SNHG9 TBC1D24 MEIOB MLST8 PRR25 FAM173A RHBDF1 CCNF SNHG19 WFIKKN1 TPSD1 LOC105371045 CACNA1H E4F1 NOXO1 PRR35 IGFALS C16orf91 MIR3176 MIR3178 EME2 PRSS29P MIR6768 LINC00254 TMEM204 SNORD60 RHBDL1 RPL3L MCRIP2 TSR3 MSLN LOC100287175 CEMP1 LINC00235 GFER HAGHL BRICD5 MPG DECR2 STUB1 TPSG1 NME4 WASIR2 GNPTG SSTR5-AS1 TMEM8A BAIAP3 RHOT2 TPSB2 CLCN7 HBA2 SNORA78 RNPS1 NPRL3 SLC9A3R2 HBQ1 ABCA17P HN1L WDR24 LOC105371184 LOC105371046 JMJD8 LOC107984876 ABCA3 RNF151 HS3ST6 LOC100134368 UBE2I ECI1 MIR662 TELO2 MIR6859-4 MIR3180-5 SPSB3 NME3 LOC107987233 TSC2 NDUFB10 POLR3K SSTR5 GNG13 SNRNP25 CRAMP1 ARHGDIG TRAF7 LMF1 MRPS34 RPS2 NTHL1 CCDC154 UNKL PGP LMF1-AS1 PDPK1 NPW MIR6767 RAB11FIP3 KCTD5 NARFL NHLRC4 LOC105371038 MIR1225 MIR3177 CAPN15 CCDC78 PKD1 RPL23AP5 FBXL16 CHTF18 AXIN1 NTN3 DDX11L10 ZNF598 SNORA10 LOC729652 PRSS27 HAGH MRPL28 METTL26 CASKIN1 LOC652276 SNORA64 AMDHD2 MAPK8IP3 LOC101929280 MIR4516 FAM234A MIR5587 PIGQ MSRB1 SYNGR3 TPSAB1 C1QTNF8 RAB26 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | HAGH CRAMP1 LOC105371047 TMEM204 FAHD1 MRPS34 TELO2 MEIOB MAPK8IP3 SPSB3 MIR3177 NME3 IFT140 IGFALS NUBP2 HN1L LOC105371046 EME2 |
nsv1046474 | Chr.16:1846666 - 1873690 on Build GRCh38 | Loss | MEIOB |
nsv519366 | Chr.16:1851210 - 1961125 on Build GRCh38 | Loss | LINC00254 HS3ST6 MSRB1 RPL3L NDUFB10 MEIOB |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | HAGH FAHD1 IGFALS MEIOB NUBP2 |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | HAGH FAHD1 MEIOB |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | HAGH LINC00254 FAHD1 MEIOB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |