Assay Details
Target Gene Details
Entrez Gene ID: | 113622 |
Gene Name: | ADP-ribosylhydrolase like 1 |
Gene Aliases: |
ARH2 |
Location: |
Chr.13:113421940-113453524 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADPRHL1 | NM_138430.4 | 7 | 1915 | NP_612439.2 |
NM_199162.2 | 7 | 1758 | NP_954631.1 | |
AA430157.1 | 1 | 126 | ||
BC009755.2 | 7 | 1758 | AAH09755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain | LOC102724474 MCF2L MIR8075 LOC105370378 GRTP1-AS1 PROZ TFDP1 ADPRHL1 LOC101928841 GRTP1 PCID2 CUL4A TMCO3 F10-AS1 LAMP1 F10 DCUN1D2 F7 |
dgv1744n100 | Chr.13:113384831 - 113442576 on Build GRCh38 | Gain | ADPRHL1 LOC101928841 |
nsv509522 | Chr.13:113408681 - 113432073 on Build GRCh38 | Insertion | ADPRHL1 LOC101928841 |
nsv563318 | Chr.13:113319297 - 113430196 on Build GRCh38 | Loss | ADPRHL1 LOC105370378 LOC101928841 GRTP1 GRTP1-AS1 LAMP1 |
nsv1195 | Chr.13:113393584 - 113427243 on Build GRCh38 | Insertion | ADPRHL1 LOC101928841 |
nsv951928 | Chr.13:113392386 - 113435785 on Build GRCh38 | Deletion | ADPRHL1 LOC101928841 |
nsv563321 | Chr.13:113369086 - 113452383 on Build GRCh38 | Loss | ADPRHL1 LOC101928841 |
esv2759970 | Chr.13:113384114 - 113556749 on Build GRCh38 | Loss | ADPRHL1 LOC101928841 TMCO3 DCUN1D2 |
nsv1044941 | Chr.13:113369130 - 113433835 on Build GRCh38 | Gain | ADPRHL1 LOC101928841 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |