Assay Details
Target Gene Details
Entrez Gene ID: | 3383 |
Gene Name: | intercellular adhesion molecule 1 |
Gene Aliases: |
BB2, CD54, P3.58 |
Location: |
Chr.19:10270841-10286615 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ICAM1 | NM_000201.2 | 7 | 1966 | NP_000192.2 |
AF340038.1 | 7 | 1647 | AAQ14901.1 | |
AF340039.1 | 7 | 1635 | AAQ14902.1 | |
AK130659.1 | 7 | 1687 | ||
AK298054.1 | 7 | 1573 | ||
AK298983.1 | 7 | 1399 | ||
AK301412.1 | 5 | 1048 | ||
AK312636.1 | ||||
BC015969.2 | 7 | 1718 | AAH15969.1 | |
BT006854.1 | AAP35500.1 | |||
J03132.1 | 7 | 1704 | AAA52709.1 | |
M24283.1 | 7 | 1718 | AAA52708.1 | |
M55038.1 | 4 | 879 | AAA60393.1 | |
X06990.1 | 7 | 1659 | CAA30051.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |