Assay Details
Target Gene Details
Entrez Gene ID: | 91304 |
Gene Name: | transmembrane protein 259 |
Gene Aliases: |
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3 |
Location: |
Chr.19:1009651-1021169 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM259 | NM_001033026.1 | 2 | 616 | NP_001028198.1 |
NM_033420.3 | 2 | 616 | NP_219488.1 | |
XM_005259675.2 | 2 | 643 | XP_005259732.1 | |
XM_005259676.2 | 2 | 643 | XP_005259733.1 | |
XM_005259677.2 | 2 | 643 | XP_005259734.1 | |
XM_005259678.2 | 2 | 643 | XP_005259735.1 | |
XM_017027457.1 | 2 | 643 | XP_016882946.1 | |
AK095756.1 | 2 | 616 | ||
BC008957.2 | 2 | 589 | AAH08957.1 | |
BQ895919.1 | 2 | 387 | ||
DQ005958.1 | 2 | 598 | AAY27747.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9662 | Chr.19:893394 - 1317740 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3643384 | Chr.19:1012803 - 1022949 on Build GRCh38 | Loss |
![]() ![]() |
nsv1160555 | Chr.19:887275 - 1346084 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3643383 | Chr.19:1012555 - 1069137 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv578188 | Chr.19:923744 - 1059005 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469845 | Chr.19:951640 - 1144486 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)