Assay Details
Target Gene Details
Entrez Gene ID: | 80333 |
Gene Name: | potassium voltage-gated channel interacting protein 4 |
Gene Aliases: |
CALP, KCHIP4 |
Location: |
Chr.4:20728520-21948801 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Target Gene Details
Entrez Gene ID: | 133015 |
Gene Name: | PARK2 coregulated like |
Gene Aliases: |
C4orf28 |
Location: |
Chr.4:20696282-20757766 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PACRGL | NR_133933.1 | |||
XM_011513786.2 | 11 | 4005 | XP_011512088.1 | |
XM_011513805.2 | 8 | 3546 | XP_011512107.1 | |
XM_017007747.1 | 10 | 3760 | XP_016863236.1 | |
XM_017007748.1 | 10 | 3711 | XP_016863237.1 | |
XM_017007749.1 | 9 | 3626 | XP_016863238.1 | |
XM_017007751.1 | 9 | 3828 | XP_016863240.1 | |
XM_017007752.1 | 8 | 3675 | XP_016863241.1 | |
XM_017007753.1 | 8 | 3545 | XP_016863242.1 | |
XM_017007754.1 | 6 | 3602 | XP_016863243.1 | |
XM_017007755.1 | 6 | 3419 | XP_016863244.1 | |
XM_017007756.1 | 6 | 3412 | XP_016863245.1 | |
XM_017007757.1 | 6 | 3348 | XP_016863246.1 | |
XM_017007758.1 | 5 | 3297 | XP_016863247.1 | |
XM_017007759.1 | 5 | 3245 | XP_016863248.1 | |
AK098692.1 | BAC05381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829876 | Chr.4:20637113 - 20833998 on Build GRCh38 | Gain | PACRGL KCNIP4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |