Assay Details
Target Gene Details
Entrez Gene ID: | 100126793 |
Gene Name: | ghrelin opposite strand/antisense RNA |
Gene Aliases: |
GHRL-AS1, GHRLAS, NCRNA00068 |
Location: |
Chr.3:10280952-10293449 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRLOS | NR_004431.3 | |||
NR_024144.2 | ||||
NR_024145.2 | ||||
NR_073566.1 | ||||
NR_073567.1 | ||||
NR_073568.1 | ||||
EU789528.1 | ||||
EU789529.1 | ||||
EU789530.1 | ||||
EU789534.1 | ||||
EU789535.1 | ||||
EU789536.1 | ||||
EU789537.1 | ||||
EU789538.1 | ||||
EU789539.1 | ||||
EU789540.1 | ||||
EU789541.1 | ||||
EU789542.1 | ||||
EU789543.1 | ||||
EU789544.1 | ||||
EU789545.1 | ||||
EU789546.1 | ||||
EU789547.1 | ||||
EU789548.1 | ||||
EU789549.1 | ||||
EU789550.1 | ||||
EU789551.1 | ||||
EU789552.1 |
Target Gene Details
Entrez Gene ID: | 84657 |
Gene Name: | long intergenic non-protein coding RNA 852 |
Gene Aliases: |
C3orf42, GHRL-AS2, GHRLOS2, NAG73 |
Location: |
Chr.3:10284419-10285746 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00852 | NR_026829.1 | 1 | 516 | |
AF280797.2 | 1 | 516 | AAK15459.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | SLC6A1-AS1 FANCD2 TAMM41 GHRL ATP2B2-IT2 FANCD2OS LINC00852 LINC00606 SEC13 LOC101927467 VGLL4 BRK1 IRAK2 VHL SLC6A1 HRH1 GHRLOS ATG7 MIR885 TATDN2 SLC6A11 ATP2B2 MIR378B |
nsv508903 | Chr.3:10275353 - 10401894 on Build GRCh38 | Insertion | GHRL LINC00852 GHRLOS MIR885 SEC13 TATDN2 ATP2B2 MIR378B |
nsv589650 | Chr.3:10260488 - 10294102 on Build GRCh38 | Loss | GHRL LINC00852 GHRLOS TATDN2 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | SLC6A1-AS1 FANCD2 GHRL ATP2B2-IT2 FANCD2OS FANCD2P2 LINC00852 SEC13 LOC101927467 EMC3 IRAK2 LOC401052 SLC6A1 HRH1 GHRLOS TATDN2 SLC6A11 ATP2B2 CIDECP TAMM41 LINC00606 VGLL4 BRK1 VHL ATG7 MIR885 MIR378B |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | SLC6A1-AS1 FANCD2 TAMM41 GHRL ATP2B2-IT2 FANCD2OS FANCD2P2 LINC00852 LINC00606 SEC13 LOC101927467 VGLL4 BRK1 IRAK2 VHL SLC6A1 HRH1 GHRLOS ATG7 MIR885 TATDN2 SLC6A11 ATP2B2 MIR378B |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | SLC6A1-AS1 FANCD2 TAMM41 GHRL ATP2B2-IT2 FANCD2OS FANCD2P2 LINC00852 LINC00606 SEC13 LOC101927467 VGLL4 BRK1 IRAK2 VHL SLC6A1 HRH1 GHRLOS ATG7 MIR885 TATDN2 SLC6A11 ATP2B2 MIR378B |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | SLC6A1-AS1 FANCD2 GHRL ATP2B2-IT2 FANCD2OS FANCD2P2 LINC00852 SEC13 LOC101927467 EMC3 IRAK2 LOC401052 SLC6A1 HRH1 GHRLOS TATDN2 SLC6A11 ATP2B2 CIDECP TAMM41 LINC00606 VGLL4 BRK1 VHL ATG7 MIR885 EMC3-AS1 MIR378B |