Assay Details
Target Gene Details
Entrez Gene ID: | 27158 |
Gene Name: | NADPH dependent diflavin oxidoreductase 1 |
Gene Aliases: |
NR1, bA350O14.9 |
Location: |
Chr.9:137205667-137219361 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDOR1 | NM_001144026.2 | NP_001137498.1 | ||
NM_001144027.2 | NP_001137499.1 | |||
NM_001144028.2 | NP_001137500.1 | |||
NM_014434.3 | NP_055249.1 | |||
XM_011518545.1 | XP_011516847.1 | |||
XM_011518547.1 | XP_011516849.1 | |||
XM_011518548.2 | XP_011516850.1 | |||
XM_017014626.1 | XP_016870115.1 | |||
XM_017014627.1 | XP_016870116.1 | |||
AF199509.1 | AAF25205.1 | |||
AK026089.1 | ||||
AK074403.1 | ||||
AK225995.1 | ||||
AK290026.1 | ||||
AY077845.1 | AAL77754.1 | |||
BC015735.1 | AAH15735.1 | |||
BC093782.1 | AAH93782.1 | |||
BC111943.1 | AAI11944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13010n54 | Chr.9:137191539 - 137233413 on Build GRCh38 | Loss | RNF208 TPRN TMEM203 NDOR1 SLC34A3 RNF224 CYSRT1 |
nsv616012 | Chr.9:137197680 - 137223834 on Build GRCh38 | Loss | RNF208 TPRN TMEM203 NDOR1 |
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss | DPP7 TMEM210 SSNA1 TPRN LOC101930307 MAN1B1 NDOR1 MAN1B1-AS1 SLC34A3 RNF224 LRRC26 RNF208 TMEM203 ANAPC2 MIR3621 UAP1L1 CYSRT1 GRIN1 |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | SSNA1 TPRN NRARP FAM166A TOR4A NDOR1 C9orf173-AS1 NELFB SLC34A3 TUBB4B RNF224 STPG3 RNF208 TMEM203 ANAPC2 CYSRT1 EXD3 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | TMEM210 PHPT1 NSMF CCDC183 LCNL1 TOR4A NDOR1 C8G C9orf139 LCN12 TRAF2 LRRC26 FUT7 MIR4479 STPG3 NPDC1 TMEM203 ANAPC2 MIR3621 FBXW5 GRIN1 ENTPD2 TPRN LOC101930307 NRARP FAM166A MAN1B1 NELFB SLC34A3 MIR7114 C9orf172 MIR4292 NOXA1 CLIC3 EDF1 RABL6 PTGDS DPP7 MAN1B1-AS1 PRR31 RNF208 ENTPD8 SAPCD2 UAP1L1 EXD3 SSNA1 C9orf173-AS1 TUBB4B RNF224 MAMDC4 CCDC183-AS1 ABCA2 PNPLA7 C9orf142 CYSRT1 |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | TMEM210 SSNA1 TPRN NRARP FAM166A TOR4A NDOR1 C9orf173-AS1 NELFB SLC34A3 TUBB4B RNF224 LRRC26 STPG3 RNF208 TMEM203 ANAPC2 MIR3621 CYSRT1 GRIN1 EXD3 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | TMEM210 SSNA1 TPRN NRARP FAM166A TOR4A NDOR1 C9orf173-AS1 NELFB SLC34A3 TUBB4B RNF224 LRRC26 STPG3 RNF208 NOXA1 TMEM203 ENTPD8 ANAPC2 MIR3621 CYSRT1 GRIN1 EXD3 |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | TMEM210 PHPT1 CCDC183 LCNL1 TOR4A NDOR1 SNHG7 C8G C9orf139 LCN12 TRAF2 SNORA17A LRRC26 FUT7 MIR4479 STPG3 NPDC1 TMEM203 ANAPC2 MIR3621 FBXW5 GRIN1 ENTPD2 EGFL7 TPRN LOC101930307 NRARP FAM166A MAN1B1 AGPAT2 NELFB SLC34A3 C9orf172 MIR4292 LCN8 CLIC3 EDF1 LCN10 RABL6 LOC100128593 PTGDS DPP7 MAN1B1-AS1 MIR6722 PRR31 RNF208 SNORA17B LCN6 TMEM141 SAPCD2 FAM69B UAP1L1 EXD3 HSPC324 SSNA1 LCN15 C9orf173-AS1 TUBB4B RNF224 MAMDC4 LINC01451 CCDC183-AS1 ABCA2 C9orf142 MIR126 CYSRT1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |