Assay Details
Target Gene Details
Entrez Gene ID: | 6929 |
Gene Name: | transcription factor 3 |
Gene Aliases: |
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21 |
Location: |
Chr.19:1609290-1652590 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | ADAMTSL5 MBD3 TCF3 MUM1 MEX3D LOC100288123 CSNK1G2 SCAMP4 ATP8B3 APC2 DAZAP1 UQCR11 C19orf25 MIR1909 KLF16 ONECUT3 CSNK1G2-AS1 RPS15 REEP6 NDUFS7 GAMT BTBD2 PCSK4 ADAT3 ABHD17A REXO1 PLK5 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | ADAMTSL5 MBD3 TCF3 MUM1 MEX3D LOC100288123 MKNK2 CSNK1G2 SCAMP4 ATP8B3 APC2 DAZAP1 UQCR11 C19orf25 MIR1909 KLF16 ONECUT3 CSNK1G2-AS1 RPS15 REEP6 NDUFS7 GAMT BTBD2 PCSK4 ADAT3 ABHD17A REXO1 PLK5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |