Assay Details
Target Gene Details
Entrez Gene ID: | 3550 |
Gene Name: | IK cytokine, down-regulator of HLA II |
Gene Aliases: |
CSA2, RED, RER |
Location: |
Chr.5:140647799-140662480 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IK | NM_006083.3 | NP_006074.2 | ||
AF182645.1 | AAD55447.1 | |||
AK225924.1 | ||||
AK293626.1 | ||||
AK308730.1 | ||||
AK312363.1 | ||||
AL050296.1 | CAB43395.2 | |||
BC013005.2 | AAH13005.1 | |||
BC071964.1 | AAH71964.1 |
Target Gene Details
Entrez Gene ID: | 100500820 |
Gene Name: | microRNA 3655 |
Gene Aliases: |
- |
Location: |
Chr.5:140647844-140647926 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIR3655 | NR_037428.1 |
Target Gene Details
Entrez Gene ID: | 4695 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A2 |
Gene Aliases: |
B8, CD14, CIB8 |
Location: |
Chr.5:140645363-140647785 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA2 | NM_001185012.1 | NP_001171941.1 | ||
NM_002488.4 | NP_002479.1 | |||
NR_033697.1 | ||||
BM706616.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |