Assay Details
Target Gene Details
Entrez Gene ID: | 56980 |
Gene Name: | PR domain 10 |
Gene Aliases: |
PFM7 |
Location: |
Chr.11:129899706-130003097 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRDM10 | NM_020228.2 | NP_064613.2 | ||
NM_199437.1 | NP_955469.1 | |||
NM_199438.1 | NP_955470.1 | |||
NM_199439.1 | NP_955471.1 | |||
XM_011542912.2 | XP_011541214.1 | |||
XM_017018023.1 | XP_016873512.1 | |||
AB033057.2 | BAA86545.2 | |||
AF275817.1 | AAF87243.1 | |||
AF503171.1 | AAP30847.1 | |||
AK000234.1 | BAA91026.1 | |||
AK310354.1 | ||||
AL833172.1 | ||||
BC112934.1 | AAI12935.1 | |||
BC117415.1 | ||||
BC143612.1 | ||||
BC143613.1 | ||||
BX648944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556564 | Chr.11:129908477 - 129934918 on Build GRCh38 | Gain | PRDM10 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |