Assay Details
Target Gene Details
Entrez Gene ID: | 25915 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 3 |
Gene Aliases: |
2P1, C3orf60, E3-3 |
Location: |
Chr.3:49020475-49023495 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAF3 | NM_199069.1 | 5 | 1060 | NP_951032.1 |
NM_199070.1 | 5 | 623 | NP_951033.1 | |
NM_199073.1 | 5 | 694 | NP_951047.1 | |
NM_199074.1 | 5 | 575 | NP_951056.1 | |
AA907148.1 | 1 | 328 | ||
AL049955.1 | 5 | 627 | CAB43224.2 | |
BC002873.2 | 5 | 610 | AAH02873.1 | |
BC111004.1 | 5 | 1690 | ||
BM985326.1 | 1 | 335 | ||
BQ652614.1 | 5 | 575 | ||
CD358942.1 | 5 | 764 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |