Assay Details
Target Gene Details
Entrez Gene ID: | 54869 |
Gene Name: | EPS8 like 1 |
Gene Aliases: |
DRC3, EPS8R1, PP10566 |
Location: |
Chr.19:55075853-55087923 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS8L1 | NM_133180.2 | NP_573441.2 | ||
XM_005259020.1 | XP_005259077.1 | |||
XM_011527051.2 | XP_011525353.1 | |||
XM_011527052.2 | XP_011525354.1 | |||
AF370395.1 | AAQ15231.1 | |||
AK075098.1 | BAC11399.1 | |||
AK296732.1 | ||||
AK309305.1 | ||||
AK309908.1 | ||||
AY074928.1 | AAL76117.1 | |||
BC004907.2 | AAH04907.2 | |||
DA452167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv817874 | Chr.19:55067334 - 55103555 on Build GRCh38 | Loss | PPP1R12C EPS8L1 RDH13 |
esv3644829 | Chr.19:55030893 - 55138732 on Build GRCh38 | Gain | PPP1R12C EPS8L1 MIR7975 GP6 RDH13 TNNT1 |
esv2667609 | Chr.19:54472514 - 55094226 on Build GRCh38 | Deletion | LILRA1 KIR2DS4 CDC42EP5 LILRP2 LILRB1 KIR3DX1 LILRA2 KIR3DL2 KIR2DL1 LOC102724300 PPP1R12C NLRP7 MIR8061 LOC101928804 KIR3DL1 KIR2DL4 LILRB4 RDH13 FCAR NLRP2 EPS8L1 NCR1 LAIR2 GP6 KIR3DL3 KIR2DL3 |
esv3893251 | Chr.19:55079303 - 55094733 on Build GRCh38 | Loss | PPP1R12C EPS8L1 |
esv2758772 | Chr.19:55026094 - 55139319 on Build GRCh38 | Gain | PPP1R12C EPS8L1 MIR7975 GP6 RDH13 TNNT1 |
nsv817875 | Chr.19:55070143 - 55090747 on Build GRCh38 | Loss | EPS8L1 |
esv2718872 | Chr.19:54472594 - 55094247 on Build GRCh38 | Deletion | LILRA1 KIR2DS4 CDC42EP5 LILRP2 LILRB1 KIR3DX1 LILRA2 KIR3DL2 KIR2DL1 LOC102724300 PPP1R12C NLRP7 MIR8061 LOC101928804 KIR3DL1 KIR2DL4 LILRB4 RDH13 FCAR NLRP2 EPS8L1 NCR1 LAIR2 GP6 KIR3DL3 KIR2DL3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |