Assay Details
Target Gene Details
Entrez Gene ID: | 2068 |
Gene Name: | ERCC excision repair 2, TFIIH core complex helicase subunit |
Gene Aliases: |
COFS2, EM9, TFIIH, TTD, TTD1, XPD |
Location: |
Chr.19:45349837-45370647 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERCC2 | NM_000400.3 | NP_000391.1 | ||
NM_001130867.1 | NP_001124339.1 | |||
XM_011526611.2 | XP_011524913.1 | |||
XM_017026467.1 | XP_016881956.1 | |||
AK303358.1 | ||||
AL535248.3 | ||||
BC108255.1 | AAI08256.1 | |||
BC110522.1 | AAI10523.1 | |||
BC110523.1 | AAI10524.1 | |||
BM769772.1 | ||||
BT006883.1 | AAP35529.1 | |||
BX376468.2 | ||||
X52221.1 | CAA36463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss | CKM ERCC2 PPP1R13L KLC3 CD3EAP MIR6088 ERCC1 |
dgv29n64 | Chr.19:45334011 - 45373709 on Build GRCh38 | Gain | ERCC2 KLC3 |
esv3644502 | Chr.19:45323147 - 45403933 on Build GRCh38 | Gain | ERCC2 PPP1R13L KLC3 |
dgv1054e212 | Chr.19:45347119 - 45407663 on Build GRCh38 | Gain | ERCC2 PPP1R13L KLC3 CD3EAP ERCC1 |
nsv458711 | Chr.19:45323108 - 45408744 on Build GRCh38 | Gain | ERCC2 PPP1R13L KLC3 CD3EAP ERCC1 |
esv3644504 | Chr.19:45342675 - 45373732 on Build GRCh38 | Gain | ERCC2 KLC3 |
nsv1063547 | Chr.19:45322368 - 45400204 on Build GRCh38 | Gain | CKM ERCC2 PPP1R13L KLC3 |
nsv833844 | Chr.19:45181051 - 45384148 on Build GRCh38 | Loss | MARK4 CKM ERCC2 PPP1R13L KLC3 EXOC3L2 BLOC1S3 |
dgv3606n100 | Chr.19:45347435 - 45400204 on Build GRCh38 | Gain | ERCC2 PPP1R13L KLC3 |
nsv428365 | Chr.19:45358027 - 45469089 on Build GRCh38 | Gain+Loss | FOSB ERCC2 PPP1R13L CD3EAP MIR6088 ERCC1 |
nsv579869 | Chr.19:45356868 - 45365051 on Build GRCh38 | Loss | ERCC2 |
nsv1160648 | Chr.19:45347447 - 45368176 on Build GRCh38 | Duplication | ERCC2 KLC3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |