Assay Details
Target Gene Details
Entrez Gene ID: | 3695 |
Gene Name: | integrin subunit beta 7 |
Gene Aliases: |
- |
Location: |
Chr.12:53191318-53207376 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGB7 | NM_000889.2 | NP_000880.1 | ||
NR_104181.1 | ||||
XM_005268851.3 | XP_005268908.1 | |||
XM_005268852.4 | XP_005268909.1 | |||
XM_006719376.3 | XP_006719439.1 | |||
AK298239.1 | ||||
AK300960.1 | ||||
BC015916.1 | AAH15916.1 | |||
M62880.1 | AAA59185.1 | |||
M68892.1 | AAA59184.1 | |||
S80335.1 | AAB21332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain | RARG CSAD IGFBP6 ZNF740 ITGB7 SPRYD3 SOAT2 MFSD5 |
nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss | RARG CSAD ESPL1 ZNF740 ITGB7 SOAT2 MFSD5 |
dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss | RARG CSAD IGFBP6 ZNF740 ITGB7 SOAT2 |
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | MIR6757 SP7 IGFBP6 LOC283335 ESPL1 ITGB7 MFSD5 RARG PFDN5 EIF4B CSAD TNS2 C12orf10 ZNF740 AAAS SPRYD3 SOAT2 |
nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss | RARG CSAD ZNF740 ITGB7 SOAT2 |
dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss | RARG CSAD TNS2 IGFBP6 ZNF740 ITGB7 SPRYD3 SOAT2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |