Assay Details
Target Gene Details
Entrez Gene ID: | 23528 |
Gene Name: | zinc finger protein 281 |
Gene Aliases: |
ZBP-99, ZNP-99 |
Location: |
Chr.1:200404947-200483604 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF281 | NM_001281293.1 | 2 | 1765 | NP_001268222.1 |
NM_001281294.1 | 3 | 1657 | NP_001268223.1 | |
NM_012482.4 | 2 | 1761 | NP_036614.1 | |
XM_017000887.1 | 5 | 3146 | XP_016856376.1 | |
XM_017000888.1 | 5 | 2104 | XP_016856377.1 | |
AF125158.1 | 1 | 1657 | AAD21084.1 | |
AJ132591.1 | 1 | 1178 | CAB70967.1 | |
AJ132592.1 | 2 | 1741 | CAB70968.1 | |
AK021707.1 | 1 | 64 | ||
AK022916.1 | 2 | 1692 | ||
AK022921.1 | 1 | 391 | ||
AK027284.1 | ||||
AK094701.1 | 3 | 1592 | ||
AK297233.1 | 3 | 1615 | ||
BC051905.1 | 1 | 1650 | AAH51905.1 | |
BC060820.1 | 2 | 1741 | AAH60820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv548990 | Chr.1:200400630 - 200452338 on Build GRCh38 | Loss | ZNF281 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |