Assay Details
Target Gene Details
Entrez Gene ID: | 2137 |
Gene Name: | exostosin like glycosyltransferase 3 |
Gene Aliases: |
BOTV, EXTL1L, EXTR1, REGR, RPR |
Location: |
Chr.8:28701473-28753690 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXTL3 | NM_001440.3 | 3 | 1872 | NP_001431.1 |
NR_073468.1 | ||||
NR_073469.1 | ||||
XM_011544440.2 | 2 | 1648 | XP_011542742.1 | |
AB007042.1 | 3 | 1709 | BAA24080.1 | |
AB011091.1 | 3 | 1731 | ||
AF001690.1 | 3 | 1717 | AAC39598.1 | |
AF029231.1 | 1 | 1378 | AAD01877.1 | |
AF083551.1 | 3 | 1709 | AAD42041.1 | |
BC006363.2 | 1 | 1228 | AAH06363.1 | |
BI911162.1 | ||||
BT007353.1 | 1 | 1116 | AAP36017.1 | |
DA222815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv1008413 | Chr.8:28716190 - 28721808 on Build GRCh38 | Deletion | EXTL3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |