Assay Details
Target Gene Details
Entrez Gene ID: | 51380 |
Gene Name: | cysteine sulfinic acid decarboxylase |
Gene Aliases: |
CSD, PCAP |
Location: |
Chr.12:53157663-53180909 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain | ZNF740 SPRYD3 CSAD RARG MFSD5 ITGB7 SOAT2 IGFBP6 |
nsv713 | Chr.12:53118449 - 53160107 on Build GRCh38 | Insertion | CSAD SOAT2 |
nsv470294 | Chr.12:53051498 - 53160499 on Build GRCh38 | Loss | TNS2 SPRYD3 CSAD MIR6757 SOAT2 LOC283335 IGFBP6 |
nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss | ZNF740 CSAD RARG MFSD5 ITGB7 SOAT2 ESPL1 |
dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss | ZNF740 CSAD RARG ITGB7 SOAT2 IGFBP6 |
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | ZNF740 TNS2 RARG PFDN5 ITGB7 SP7 AAAS EIF4B MIR6757 C12orf10 SPRYD3 CSAD MFSD5 SOAT2 LOC283335 ESPL1 IGFBP6 |
nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss | ZNF740 CSAD RARG ITGB7 SOAT2 |
dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss | ZNF740 TNS2 SPRYD3 CSAD RARG ITGB7 SOAT2 IGFBP6 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |