Assay Details
Target Gene Details
Entrez Gene ID: | 8372 |
Gene Name: | hyaluronoglucosaminidase 3 |
Gene Aliases: |
HYAL-3, LUCA-3, LUCA3 |
Location: |
Chr.3:50292828-50299468 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYAL3 | NM_001200029.1 | 4 | 1354 | NP_001186958.1 |
NM_001200030.1 | 3 | 1478 | NP_001186959.1 | |
NM_001200031.1 | 4 | 802 | NP_001186960.1 | |
NM_001200032.1 | 3 | 712 | NP_001186961.1 | |
NM_003549.3 | 4 | 1568 | NP_003540.2 | |
AF036035.1 | 4 | 1568 | AAD04257.1 | |
AF040710.1 | 4 | 1488 | AAC70915.1 | |
AF502909.1 | 3 | 1242 | AAM60775.1 | |
AF502910.1 | 4 | 566 | AAM60776.1 | |
AF502911.1 | 3 | 476 | AAM60777.1 | |
AF502912.1 | 4 | 1332 | AAM60778.1 | |
AK313455.1 | ||||
BC005896.1 | 4 | 1458 | AAH05896.1 | |
BC012892.1 | 3 | 1323 | AAH12892.1 | |
DB319349.1 | 1 | 378 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759148 | Chr.3:50161045 - 50333946 on Build GRCh38 | Loss | HYAL3 SLC38A3 LSMEM2 RASSF1 MIR6872 HYAL1 TUSC2 IFRD2 MIR566 SEMA3F SEMA3B-AS1 NAT6 HYAL2 GNAI2 MIR5787 SEMA3B GNAT1 |
nsv523560 | Chr.3:50134964 - 50314769 on Build GRCh38 | Loss | HYAL3 SLC38A3 LSMEM2 MIR6872 HYAL1 IFRD2 MIR566 LOC107986035 SEMA3F-AS1 SEMA3F SEMA3B-AS1 NAT6 GNAI2 MIR5787 SEMA3B GNAT1 |
dgv8317n54 | Chr.3:50206077 - 50383123 on Build GRCh38 | Loss | HYAL3 SLC38A3 RASSF1-AS1 LSMEM2 RASSF1 NPRL2 MIR6872 TMEM115 CYB561D2 HYAL1 TUSC2 IFRD2 ZMYND10 SEMA3B-AS1 NAT6 HYAL2 GNAI2 MIR5787 CACNA2D2 SEMA3B |
nsv834693 | Chr.3:50251400 - 50470533 on Build GRCh38 | Loss | HYAL3 RASSF1-AS1 LSMEM2 RASSF1 NPRL2 MIR6872 TMEM115 CYB561D2 HYAL1 TUSC2 IFRD2 ZMYND10 SEMA3B-AS1 NAT6 HYAL2 GNAI2 CACNA2D2 SEMA3B |
nsv834692 | Chr.3:50161052 - 50356033 on Build GRCh38 | Loss | HYAL3 SLC38A3 RASSF1-AS1 LSMEM2 RASSF1 NPRL2 MIR6872 TMEM115 CYB561D2 HYAL1 TUSC2 IFRD2 MIR566 ZMYND10 SEMA3F SEMA3B-AS1 NAT6 HYAL2 GNAI2 MIR5787 SEMA3B GNAT1 |
dgv8316n54 | Chr.3:50115923 - 50299991 on Build GRCh38 | Loss | HYAL3 SLC38A3 LSMEM2 MIR6872 RBM5 HYAL1 IFRD2 MIR566 LOC107986035 SEMA3F-AS1 SEMA3F SEMA3B-AS1 NAT6 GNAI2 MIR5787 SEMA3B GNAT1 |
nsv470561 | Chr.3:50121341 - 50547568 on Build GRCh38 | Loss | SLC38A3 RASSF1-AS1 LSMEM2 RASSF1 MIR6872 TMEM115 CYB561D2 HYAL1 TUSC2 MIR566 ZMYND10 LOC107986035 SEMA3F-AS1 SEMA3F SEMA3B-AS1 SEMA3B HYAL3 NPRL2 IFRD2 NAT6 HYAL2 GNAI2 MIR5787 CACNA2D2 GNAT1 |
nsv523662 | Chr.3:50107518 - 50547568 on Build GRCh38 | Loss | SLC38A3 RASSF1-AS1 LSMEM2 RASSF1 MIR6872 TMEM115 CYB561D2 HYAL1 TUSC2 MIR566 ZMYND10 LOC107986035 SEMA3F-AS1 SEMA3F SEMA3B-AS1 SEMA3B HYAL3 NPRL2 RBM5 IFRD2 NAT6 HYAL2 GNAI2 MIR5787 CACNA2D2 GNAT1 |
nsv590255 | Chr.3:50210391 - 50399396 on Build GRCh38 | Loss | HYAL3 SLC38A3 RASSF1-AS1 LSMEM2 RASSF1 NPRL2 MIR6872 TMEM115 CYB561D2 HYAL1 TUSC2 IFRD2 ZMYND10 SEMA3B-AS1 NAT6 HYAL2 GNAI2 MIR5787 CACNA2D2 SEMA3B |
nsv954855 | Chr.3:50152468 - 50439169 on Build GRCh38 | Deletion | SLC38A3 RASSF1-AS1 LSMEM2 RASSF1 MIR6872 TMEM115 CYB561D2 HYAL1 TUSC2 MIR566 ZMYND10 LOC107986035 SEMA3F-AS1 SEMA3F SEMA3B-AS1 SEMA3B HYAL3 NPRL2 IFRD2 NAT6 HYAL2 GNAI2 MIR5787 CACNA2D2 GNAT1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |