Assay Details
Target Gene Details
Entrez Gene ID: | 49856 |
Gene Name: | WD repeat containing, antisense to TP73 |
Gene Aliases: |
WDR8 |
Location: |
Chr.1:3630767-3650107 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRAP73 | NM_017818.3 | NP_060288.3 | ||
XM_005244754.1 | XP_005244811.1 | |||
XM_017001387.1 | XP_016856876.1 | |||
AB034912.1 | BAA92312.1 | |||
AI376011.1 | ||||
AK000437.1 | BAA91164.1 | |||
AK225525.1 | ||||
BC086311.1 | AAH86311.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758915 | Chr.1:3372374 - 3632289 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv545128 | Chr.1:3630371 - 3700037 on Build GRCh38 | Loss |
![]() ![]() |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3585044 | Chr.1:3611044 - 3637348 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3585045 | Chr.1:3611161 - 3686920 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)