Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | 3 | 298 | BAC04000.1 | |
AK292004.1 | ||||
AK300915.1 | ||||
DA945486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | EXOC3L1 TPPP3 GFOD2 CENPT ZDHHC1 KCTD19 NOL3 CARMIL2 EDC4 NUTF2 FHOD1 KIAA0895L ELMO3 DPEP3 TMEM208 NRN1L RANBP10 LOC105369155 SLC12A4 LOC100131303 SLC9A5 LOC107984813 DDX28 TRADD PLEKHG4 LRRC36 MIR328 PSKH1 LRRC29 HSD11B2 FBXL8 THAP11 DPEP2 CTCF HSF4 LCAT C16orf86 TSNAXIP1 PARD6A PSMB10 DUS2 LOC100505942 ATP6V0D1 AGRP ACD FAM65A ENKD1 E2F4 CTRL |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | EXOC3L1 FHOD1 TRADD KIAA0895L ELMO3 PLEKHG4 MIR328 C16orf70 TMEM208 NOL3 LOC105369155 LRRC29 B3GNT9 FBXL8 SLC9A5 HSF4 E2F4 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | EXOC3L1 FHOD1 TRADD KIAA0895L ELMO3 MIR328 C16orf70 TMEM208 NOL3 LOC105369155 LRRC29 B3GNT9 FBXL8 HSF4 E2F4 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | LOC105369155 LRRC29 EXOC3L1 KIAA0895L ELMO3 FBXL8 MIR328 NOL3 HSF4 E2F4 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | LOC105369155 LRRC29 EXOC3L1 FHOD1 KIAA0895L ELMO3 PLEKHG4 MIR328 TMEM208 SLC9A5 E2F4 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | EXOC3L1 FHOD1 KIAA0895L ELMO3 PLEKHG4 MIR328 TMEM208 KCTD19 NOL3 LOC105369155 LRRC29 SLC9A5 E2F4 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | EXOC3L1 TPPP3 GFOD2 CENPT ZDHHC1 KCTD19 NOL3 CARMIL2 EDC4 NUTF2 FHOD1 KIAA0895L ELMO3 DPEP3 TMEM208 NRN1L RANBP10 LOC105369155 SLC12A4 LOC100131303 SLC9A5 LOC107984813 DDX28 TRADD PLEKHG4 LRRC36 MIR328 PSKH1 LRRC29 HSD11B2 B3GNT9 FBXL8 THAP11 DPEP2 CTCF HSF4 LCAT C16orf86 TSNAXIP1 PARD6A PSMB10 C16orf70 DUS2 LOC100505942 ATP6V0D1 AGRP ACD FAM65A ENKD1 E2F4 CTRL |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |