Assay Details
Target Gene Details
Entrez Gene ID: | 55911 |
Gene Name: | apolipoprotein B receptor |
Gene Aliases: |
APOB100R, APOB48R |
Location: |
Chr.16:28494649-28498970 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBR | NM_018690.3 | 3 | 3076 | NP_061160.3 |
AF141332.1 | 4 | 3026 | AAF76255.1 | |
AF141334.1 | 4 | 3026 | AAF78765.1 | |
AK025123.1 | 2 | 928 | ||
AK075085.1 | 2 | 751 | ||
BC119786.1 | 4 | 3026 | ||
BC119788.1 | 4 | 2821 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | SULT1A1 IL27 APOBR SULT1A2 CLN3 NUPR1 SGF29 |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | NPIPB8 SULT1A2 CLN3 TUFM MIR4721 XPO6 SH2B1 ATXN2L MIR6862-1 RABEP2 ATP2A1 NPIPB6 RRN3P2 SBK1 NFATC2IP APOBR NPIP NUPR1 CD19 EIF3C MIR6862-2 LOC101928215 SULT1A1 IL27 ATP2A1-AS1 SPNS1 MIR4517 EIF3CL NPIPB9 LAT SGF29 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | NPIPB8 SULT1A2 CLN3 TUFM MIR4721 LOC107984832 SH2B1 ATXN2L MIR6862-1 RABEP2 ATP2A1 RRN3P2 NFATC2IP APOBR NPIP NUPR1 CD19 EIF3C MIR6862-2 LOC101928215 SULT1A1 IL27 ATP2A1-AS1 SPNS1 MIR4517 EIF3CL NPIPB9 LAT SGF29 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | IL27 APOBR CLN3 MIR6862-1 NUPR1 EIF3CL NPIPB6 SGF29 |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | APOBR CLN3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |