Assay Details
Target Gene Details
Entrez Gene ID: | 3684 |
Gene Name: | integrin subunit alpha M |
Gene Aliases: |
CD11B, CR3A, MAC-1, MAC1A, MO1A, SLEB6 |
Location: |
Chr.16:31259967-31332892 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGAM | NM_000632.3 | 11 | 1223 | NP_000623.2 |
NM_001145808.1 | 11 | 1223 | NP_001139280.1 | |
XM_006721045.1 | 11 | 1215 | XP_006721108.1 | |
XM_011545850.2 | 9 | 1019 | XP_011544152.1 | |
XM_011545851.2 | 11 | 1215 | XP_011544153.1 | |
XM_017023216.1 | 11 | 1215 | XP_016878705.1 | |
AK057856.1 | ||||
AK127637.1 | ||||
AY187247.1 | 3 | 224 | ||
AY187248.1 | 3 | 224 | ||
BC096346.3 | 11 | 1189 | AAH96346.1 | |
BC096347.3 | 11 | 1189 | AAH96347.1 | |
BC096348.3 | 11 | 1189 | AAH96348.1 | |
BC099660.3 | 11 | 1189 | AAH99660.1 | |
J03925.1 | 11 | 1197 | AAA59544.1 | |
J04145.1 | 10 | 1101 | AAA59903.1 | |
M18044.1 | 11 | 1200 | AAA59491.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |