Assay Details
Target Gene Details
Entrez Gene ID: | 10087 |
Gene Name: | collagen type IV alpha 3 binding protein |
Gene Aliases: |
CERT, CERTL, GPBP, MRD34, STARD11 |
Location: |
Chr.5:75368486-75511981 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COL4A3BP | NM_001130105.1 | 1 | 74 | NP_001123577.1 |
AK096854.1 | 1 | 231 |
Target Gene Details
Entrez Gene ID: | 51426 |
Gene Name: | polymerase (DNA) kappa |
Gene Aliases: |
DINB1, DINP, POLQ |
Location: |
Chr.5:75511832-75602576 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLK | NM_016218.2 | NP_057302.1 | ||
XM_005248534.4 | XP_005248591.1 | |||
XM_011543464.2 | XP_011541766.1 | |||
XM_017009561.1 | XP_016865050.1 | |||
XM_017009562.1 | XP_016865051.1 | |||
XM_017009564.1 | XP_016865053.1 | |||
AF163570.1 | AAF02540.1 | |||
AF194973.1 | AAF23270.1 | |||
AK091659.1 | BAC03714.1 | |||
AK314610.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |