Assay Details
Target Gene Details
Entrez Gene ID: | 4547 |
Gene Name: | microsomal triglyceride transfer protein |
Gene Aliases: |
ABL, MTP |
Location: |
Chr.4:99564078-99623997 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTTP | NM_000253.3 | NP_000244.2 | ||
NM_001300785.1 | NP_001287714.1 | |||
AK225328.1 | ||||
AK290793.1 | ||||
AK302602.1 | ||||
BC062696.1 | AAH62696.1 | |||
BC125110.1 | ||||
BC125111.1 | ||||
DB083001.1 | ||||
DB458800.2 | 4 | 517 | ||
X59657.1 | CAA42200.1 | |||
X75500.1 | CAA53217.1 | |||
X91148.1 | CAA62585.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs141736123] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Non-exonic non-DGV Variation |