Assay Details
Target Gene Details
Entrez Gene ID: | 29882 |
Gene Name: | anaphase promoting complex subunit 2 |
Gene Aliases: |
APC2 |
Location: |
Chr.9:137174784-137188605 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANAPC2 | NM_013366.3 | NP_037498.1 | ||
AB037827.1 | BAA92644.1 | |||
AF191337.1 | AAF05751.1 | |||
AK294601.1 | ||||
BC001579.1 | AAH01579.1 | |||
BC009487.2 | AAH09487.2 | |||
BC032503.1 | AAH32503.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss | LOC101930307 CYSRT1 SSNA1 TPRN RNF224 NDOR1 SLC34A3 GRIN1 TMEM210 TMEM203 LRRC26 DPP7 MAN1B1 MIR3621 RNF208 ANAPC2 UAP1L1 MAN1B1-AS1 |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | FAM166A CYSRT1 SSNA1 NELFB TPRN RNF224 NDOR1 EXD3 SLC34A3 TOR4A TMEM203 STPG3 RNF208 NRARP ANAPC2 TUBB4B C9orf173-AS1 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | LOC101930307 PRR31 MIR7114 MAMDC4 EXD3 NPDC1 GRIN1 ABCA2 CCDC183 STPG3 C8G NRARP C9orf142 ANAPC2 PHPT1 MIR4479 UAP1L1 LCN12 NSMF CYSRT1 ENTPD2 FUT7 CLIC3 PTGDS C9orf139 PNPLA7 NDOR1 SLC34A3 TMEM210 MAN1B1 EDF1 RABL6 SAPCD2 C9orf172 TRAF2 ENTPD8 TMEM203 DPP7 CCDC183-AS1 LCNL1 FBXW5 MAN1B1-AS1 FAM166A SSNA1 NELFB NOXA1 TPRN RNF224 MIR4292 TOR4A LRRC26 MIR3621 RNF208 TUBB4B C9orf173-AS1 |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | FAM166A CYSRT1 SSNA1 NELFB TPRN RNF224 NDOR1 EXD3 SLC34A3 GRIN1 TMEM210 TOR4A TMEM203 LRRC26 STPG3 MIR3621 RNF208 NRARP ANAPC2 TUBB4B C9orf173-AS1 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | FAM166A CYSRT1 SSNA1 NELFB NOXA1 TPRN ENTPD8 RNF224 NDOR1 EXD3 SLC34A3 GRIN1 TMEM210 TOR4A TMEM203 LRRC26 STPG3 MIR3621 RNF208 NRARP ANAPC2 TUBB4B C9orf173-AS1 |
esv2761561 | Chr.9:137030185 - 137191593 on Build GRCh38 | Gain | LOC101930307 ENTPD2 SAPCD2 SSNA1 FUT7 C9orf139 NPDC1 GRIN1 TMEM210 LRRC26 DPP7 MAN1B1 MIR3621 ANAPC2 UAP1L1 MAN1B1-AS1 |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | LOC101930307 PRR31 EGFL7 LCN6 LOC100128593 LINC01451 MAMDC4 EXD3 NPDC1 GRIN1 ABCA2 CCDC183 LCN15 STPG3 C8G NRARP C9orf142 ANAPC2 PHPT1 MIR4479 UAP1L1 LCN12 MIR6722 SNORA17B CYSRT1 ENTPD2 FUT7 CLIC3 PTGDS AGPAT2 C9orf139 NDOR1 SLC34A3 HSPC324 TMEM210 MAN1B1 SNORA17A EDF1 RABL6 LCN8 TMEM141 SAPCD2 C9orf172 TRAF2 TMEM203 DPP7 LCN10 CCDC183-AS1 LCNL1 FBXW5 MAN1B1-AS1 FAM166A FAM69B SSNA1 NELFB SNHG7 TPRN RNF224 MIR4292 TOR4A LRRC26 MIR3621 RNF208 TUBB4B C9orf173-AS1 MIR126 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |