Assay Details
Target Gene Details
Entrez Gene ID: | 401431 |
Gene Name: | ATP6V0E2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:149867694-149873862 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V0E2-AS1 | NR_027040.1 | 4 | 1053 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv8229 | Chr.7:149852698 - 150185474 on Build GRCh38 | Gain+Loss | LOC100134040 ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C |
nsv1033095 | Chr.7:149810995 - 150084783 on Build GRCh38 | Gain | LOC100134040 ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C SSPO |
nsv521861 | Chr.7:149761534 - 149885817 on Build GRCh38 | Loss | ZNF862 ATP6V0E2 ATP6V0E2-AS1 ZNF467 ACTR3C SSPO |
esv2759571 | Chr.7:149744501 - 150259326 on Build GRCh38 | Gain | LOC100134040 ZNF862 ATP6V0E2 ATP6V0E2-AS1 ZNF467 ACTR3C SSPO |
esv3615438 | Chr.7:149790046 - 150039200 on Build GRCh38 | Gain | LOC100134040 ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C SSPO |
esv3576480 | Chr.7:149621871 - 150131598 on Build GRCh38 | Gain | LOC100134040 ZNF862 ATP6V0E2 ATP6V0E2-AS1 ZNF767P ZNF467 ACTR3C KRBA1 SSPO |
nsv831184 | Chr.7:149727744 - 149903004 on Build GRCh38 | Gain | ZNF862 ATP6V0E2 ATP6V0E2-AS1 ZNF467 ACTR3C KRBA1 SSPO |
esv3891234 | Chr.7:149854904 - 150009836 on Build GRCh38 | Gain | ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C |
nsv1029484 | Chr.7:149842825 - 149902237 on Build GRCh38 | Gain | ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C |
nsv1161666 | Chr.7:149855457 - 150116613 on Build GRCh38 | Duplication | LOC100134040 ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C |
nsv831185 | Chr.7:149864016 - 150022570 on Build GRCh38 | Gain | ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C |
dgv6751n100 | Chr.7:149855445 - 149998108 on Build GRCh38 | Gain | ZNF862 ATP6V0E2 ATP6V0E2-AS1 ACTR3C |