Assay Details
Target Gene Details
Entrez Gene ID: | 7163 |
Gene Name: | tumor protein D52 |
Gene Aliases: |
D52, N8L, PC-1, PrLZ, hD52 |
Location: |
Chr.8:80034868-80171659 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPD52 | NM_001025252.2 | 6 | 991 | NP_001020423.1 |
NM_001025253.2 | 8 | 778 | NP_001020424.1 | |
NM_001287140.1 | 8 | 1060 | NP_001274069.1 | |
NM_001287142.1 | 7 | 1033 | NP_001274071.1 | |
NM_001287143.1 | 7 | 1018 | NP_001274072.1 | |
NM_001287144.1 | 6 | 702 | NP_001274073.1 | |
NM_005079.3 | 6 | 709 | NP_005070.1 | |
NR_105033.1 | 9 | 1130 | ||
NR_105034.1 | 5 | 593 | ||
NR_105035.1 | 7 | 779 | ||
NR_105036.1 | 8 | 723 | ||
NR_105037.1 | 7 | 724 | ||
AF202897.1 | 6 | 935 | AAO22156.1 | |
AK057075.1 | 8 | 779 | ||
AK223631.1 | 6 | 608 | BAD97351.1 | |
AK295468.1 | 8 | 645 | ||
AK296615.1 | 6 | 713 | ||
AK308983.1 | 8 | 635 | ||
AK313135.1 | 6 | 553 | ||
BC018117.1 | 6 | 580 | AAH18117.1 | |
BI546831.1 | 8 | 680 | ||
BI551254.1 | 8 | 770 | ||
BT019444.1 | 6 | 493 | AAV38251.1 | |
BX640835.1 | 6 | 775 | CAE45908.1 | |
CR542034.1 | 7 | 493 | CAG46831.1 | |
CR542064.1 | 6 | 493 | CAG46861.1 | |
DA199944.1 | ||||
DA409017.1 | 6 | 585 | ||
DA921955.1 | ||||
DB505575.1 | ||||
GQ499324.1 | 8 | 1045 | ||
GQ499325.1 | 7 | 1018 | ||
GQ499326.1 | 7 | 1003 | ||
GQ499327.1 | 6 | 976 | ||
GQ499328.1 | 9 | 1115 | ||
S82081.1 | 6 | 586 | AAB36475.1 | |
U18914.1 | 6 | 584 | AAC50183.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |