Assay Details
Target Gene Details
Entrez Gene ID: | 201254 |
Gene Name: | stimulated by retinoic acid 13 |
Gene Aliases: |
CENP-X, CENPX, D9, FAAP10, MHF2 |
Location: |
Chr.17:82018703-82022930 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STRA13 | NM_001271006.1 | 5 | 365 | NP_001257935.1 |
NM_001271007.1 | 4 | 296 | NP_001257936.1 | |
NM_144998.3 | 4 | 311 | NP_659435.2 | |
XM_005256339.2 | 5 | 517 | XP_005256396.1 | |
XM_017024326.1 | 3 | 718 | XP_016879815.1 | |
XM_017024327.1 | 4 | 590 | XP_016879816.1 | |
XM_017024328.1 | 4 | 459 | XP_016879817.1 | |
XM_017024329.1 | 3 | 405 | XP_016879818.1 | |
BC009571.1 | 4 | 280 | AAH09571.1 | |
BC011610.1 | 4 | 247 | AAH11610.1 | |
BI837753.1 | 4 | 279 | ||
BQ648002.1 | ||||
CB110145.1 | 5 | 374 | ||
HY110543.1 | 5 | 355 | ||
U95006.1 | 4 | 266 | AAB53638.1 | |
U95007.1 | 4 | 251 | AAB53639.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952392 | Chr.17:81885825 - 82065724 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576183 | Chr.17:81991355 - 82086850 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv5800n54 | Chr.17:82013189 - 82035336 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)