Assay Details
Target Gene Details
Entrez Gene ID: | 221264 |
Gene Name: | adenylate kinase 9 |
Gene Aliases: |
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1 |
Location: |
Chr.6:109492855-109691253 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AK9 | NM_001145128.2 | NP_001138600.2 | ||
NM_145025.4 | NP_659462.1 | |||
XM_006715376.3 | XP_006715439.2 | |||
XM_011535550.2 | XP_011533852.1 | |||
XM_011535552.2 | XP_011533854.1 | |||
XM_011535553.2 | XP_011533855.1 | |||
XM_011535554.2 | XP_011533856.1 | |||
XM_011535555.2 | XP_011533857.1 | |||
XM_011535556.2 | XP_011533858.1 | |||
XM_011535559.2 | XP_011533861.1 | |||
XM_017010384.1 | XP_016865873.1 | |||
XM_017010385.1 | XP_016865874.1 | |||
XM_017010386.1 | XP_016865875.1 | |||
XM_017010388.1 | XP_016865877.1 | |||
AK055538.1 | BAB70945.1 | |||
AK315561.1 | ||||
AL832162.1 | ||||
BC022031.2 | AAH22031.1 |
Target Gene Details
Entrez Gene ID: | 9896 |
Gene Name: | FIG4 phosphoinositide 5-phosphatase |
Gene Aliases: |
ALS11, BTOP, CMT4J, KIAA0274, SAC3, YVS, dJ249I4.1 |
Location: |
Chr.6:109691216-109825431 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FIG4 | NM_014845.5 | NP_055660.1 | ||
XM_017011591.1 | XP_016867080.1 | |||
DA811740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830769 | Chr.6:109623131 - 109756917 on Build GRCh38 | Gain | AK9 FIG4 |
nsv604485 | Chr.6:109638066 - 109733283 on Build GRCh38 | Loss | AK9 FIG4 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |