Assay Details
Target Gene Details
Entrez Gene ID: | 81928 |
Gene Name: | Cdk5 and Abl enzyme substrate 2 |
Gene Aliases: |
C20orf150, dJ908M14.2, ik3-2 |
Location: |
Chr.20:62388630-62407283 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABLES2 | NM_031215.2 | 10 | 1600 | NP_112492.2 |
BC003122.1 | 8 | 1135 | AAH03122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422398 | Chr.20:62207833 - 62674040 on Build GRCh38 | Duplication | LAMA5 MIR133A2 SLCO4A1 ADRM1 GATA5 MIR4758 LOC105369209 MIR1-1 MIR1-1HG SLCO4A1-AS1 C20orf166-AS1 LAMA5-AS1 HRH3 RPS21 CABLES2 RBBP8NL OSBPL2 |
nsv828774 | Chr.20:62378159 - 62404579 on Build GRCh38 | Loss | RPS21 CABLES2 |
nsv586496 | Chr.20:62332390 - 62812195 on Build GRCh38 | Gain | LAMA5 LOC107985428 MIR133A2 SLCO4A1 GATA5 OGFR-AS1 MIR4758 OGFR MIR1-1 MIR1-1HG NTSR1 SLCO4A1-AS1 C20orf166-AS1 MRGBP LINC00659 LOC107987285 LAMA5-AS1 RPS21 CABLES2 RBBP8NL |
nsv470557 | Chr.20:62312525 - 62415019 on Build GRCh38 | Gain | LAMA5 LAMA5-AS1 MIR4758 RPS21 CABLES2 RBBP8NL |
nsv586497 | Chr.20:62345988 - 62415019 on Build GRCh38 | Loss | LAMA5 LAMA5-AS1 RPS21 CABLES2 RBBP8NL |
nsv953302 | Chr.20:62345345 - 62831248 on Build GRCh38 | Deletion | LAMA5 LOC107985428 MIR133A2 SLCO4A1 GATA5 OGFR-AS1 OGFR MIR1-1 MIR1-1HG NTSR1 SLCO4A1-AS1 C20orf166-AS1 MRGBP LINC00659 LOC107987285 LAMA5-AS1 RPS21 CABLES2 COL9A3 RBBP8NL |
nsv518296 | Chr.20:62203245 - 62415019 on Build GRCh38 | Loss | LAMA5 ADRM1 MTG2 LAMA5-AS1 MIR4758 HRH3 RPS21 CABLES2 LOC105369209 RBBP8NL OSBPL2 |
dgv7650n54 | Chr.20:62315508 - 62394547 on Build GRCh38 | Loss | LAMA5 LAMA5-AS1 MIR4758 RPS21 CABLES2 |
nsv586498 | Chr.20:62387568 - 62415019 on Build GRCh38 | Loss | RPS21 CABLES2 RBBP8NL |
nsv834024 | Chr.20:62259914 - 62455522 on Build GRCh38 | Gain | LAMA5 ADRM1 LAMA5-AS1 MIR4758 RPS21 CABLES2 RBBP8NL OSBPL2 |
nsv470556 | Chr.20:62212004 - 62577283 on Build GRCh38 | Loss | LAMA5 MIR133A2 ADRM1 GATA5 MIR4758 LOC105369209 MIR1-1 MIR1-1HG C20orf166-AS1 LAMA5-AS1 HRH3 RPS21 CABLES2 RBBP8NL OSBPL2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |