Assay Details
Target Gene Details
Entrez Gene ID: | 51654 |
Gene Name: | CDK5 regulatory subunit associated protein 1 |
Gene Aliases: |
C20orf34, C42, CGI-05, HSPC167 |
Location: |
Chr.20:33358839-33402768 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDK5RAP1 | NM_001278167.1 | 11 | 1501 | NP_001265096.1 |
NM_001278168.1 | 10 | 1270 | NP_001265097.1 | |
NM_001278169.1 | 11 | 1269 | NP_001265098.1 | |
NM_016082.4 | 11 | 1353 | NP_057166.4 | |
NM_016408.3 | 11 | 1501 | NP_057492.2 | |
XM_011528856.2 | 8 | 2237 | XP_011527158.1 | |
XM_011528857.2 | 10 | 1113 | XP_011527159.1 | |
XM_011528858.1 | 10 | 1352 | XP_011527160.1 | |
XM_011528859.2 | 10 | 1112 | XP_011527161.1 | |
XM_011528860.1 | 8 | 812 | XP_011527162.1 | |
XM_017027876.1 | 10 | 1251 | XP_016883365.1 | |
XM_017027877.1 | 5 | 565 | XP_016883366.1 | |
AF152097.1 | 12 | 1479 | AAD34147.1 | |
AF161516.1 | 11 | 1449 | AAF29131.1 | |
AK023992.1 | 11 | 1451 | BAB14760.1 | |
AK027449.1 | 11 | 1301 | BAB55120.1 | |
AK095644.1 | 2 | 801 | ||
AK222745.1 | 11 | 1451 | BAD96465.1 | |
AK292075.1 | 11 | 1219 | ||
AK295342.1 | 11 | 1437 | ||
AK314532.1 | 11 | 1451 | ||
AY462283.1 | 11 | 1452 | AAS18317.1 | |
AY462284.1 | AAS18318.1 | |||
BC001215.1 | 10 | 1199 | AAH01215.1 | |
BC050706.1 | 11 | 1444 | AAH50706.1 | |
BM792566.1 | 3 | 193 | ||
CX785345.1 | 6 | 708 | ||
KU178644.1 | 9 | 1108 | ||
KU178645.1 | 10 | 1339 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064413 | Chr.20:33348381 - 33599978 on Build GRCh38 | Gain | SNTA1 CBFA2T2 CDK5RAP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |