Assay Details
Target Gene Details
Entrez Gene ID: | 90627 |
Gene Name: | StAR related lipid transfer domain containing 13 |
Gene Aliases: |
ARHGAP37, DLC2, GT650, LINC00464 |
Location: |
Chr.13:33103135-33676835 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD13 | NM_001243476.2 | 17 | 3611 | NP_001230405.1 |
NM_052851.2 | 13 | 3201 | NP_443083.1 | |
NM_178006.3 | 13 | 3264 | NP_821074.1 | |
NM_178007.2 | 13 | 3171 | NP_821075.1 | |
XM_011535299.2 | 16 | 3357 | XP_011533601.1 | |
XM_017020834.1 | 13 | 3369 | XP_016876323.1 | |
XM_017020835.1 | 15 | 3290 | XP_016876324.1 | |
AK094704.1 | 12 | 2162 | ||
AK095114.1 | 9 | 1884 | ||
AK312888.1 | 13 | 3257 | ||
AL049801.1 | 13 | 3157 | CAB42562.1 | |
AY082589.1 | 13 | 3259 | AAL91648.1 | |
AY082590.1 | 13 | 3183 | AAL91649.1 | |
AY082591.1 | 13 | 3157 | AAL91650.1 | |
AY366448.1 | 13 | 3199 | AAQ72791.1 | |
BX641149.1 | 3 | 284 | CAE46065.1 | |
BX647695.1 | 17 | 3571 | CAI46026.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |