Assay Details
Target Gene Details
Entrez Gene ID: | 10018 |
Gene Name: | BCL2 like 11 |
Gene Aliases: |
BAM, BIM, BOD |
Location: |
Chr.2:111120914-111168445 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss | FBLN7 TMEM87B MIR4435-2 ACOXL LOC400997 BCL2L11 MIR4771-2 MERTK MIR4435-2HG ANAPC1 PAFAH1B1P2 |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | FBLN7 ZC3H6 MIR4771-2 TMEM87B BUB1 SNORD132 MIR4435-2 ACOXL LOC400997 BCL2L11 MERTK MIR4435-2HG ANAPC1 ZC3H8 PAFAH1B1P2 |
nsv834331 | Chr.2:111054562 - 111247101 on Build GRCh38 | Loss | ACOXL LOC400997 BCL2L11 MIR4435-2HG |
nsv428402 | Chr.2:111142206 - 111905020 on Build GRCh38 | Loss | MIR4435-2 BCL2L11 MIR4771-2 MERTK MIR4435-2HG ANAPC1 PAFAH1B1P2 |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | LOC105375809 LIMS4 FBLN7 RGPD6 LINC01106 ZC3H6 LOC100288570 MIR4771-2 TMEM87B BUB1 SNORD132 MIR4435-2 ACOXL LOC105373553 LOC400997 BCL2L11 MERTK MIR4435-2HG ANAPC1 ZC3H8 PAFAH1B1P2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |