Assay Details
Target Gene Details
Entrez Gene ID: | 81928 |
Gene Name: | Cdk5 and Abl enzyme substrate 2 |
Gene Aliases: |
C20orf150, dJ908M14.2, ik3-2 |
Location: |
Chr.20:62388630-62407283 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABLES2 | NM_031215.2 | NP_112492.2 | ||
BC003122.1 | AAH03122.1 | |||
BU166894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422398 | Chr.20:62207833 - 62674040 on Build GRCh38 | Duplication | LOC105369209 LAMA5-AS1 ADRM1 CABLES2 SLCO4A1-AS1 LAMA5 MIR1-1HG SLCO4A1 C20orf166-AS1 OSBPL2 RPS21 RBBP8NL MIR133A2 GATA5 MIR1-1 MIR4758 HRH3 |
nsv828774 | Chr.20:62378159 - 62404579 on Build GRCh38 | Loss | RPS21 CABLES2 |
nsv586496 | Chr.20:62332390 - 62812195 on Build GRCh38 | Gain | LOC107987285 LAMA5-AS1 LINC00659 OGFR-AS1 CABLES2 SLCO4A1-AS1 LAMA5 MIR1-1HG LOC107985428 SLCO4A1 C20orf166-AS1 OGFR NTSR1 RPS21 RBBP8NL MIR133A2 GATA5 MIR1-1 MIR4758 MRGBP |
nsv470557 | Chr.20:62312525 - 62415019 on Build GRCh38 | Gain | LAMA5-AS1 RPS21 RBBP8NL CABLES2 LAMA5 MIR4758 |
nsv586497 | Chr.20:62345988 - 62415019 on Build GRCh38 | Loss | LAMA5-AS1 RPS21 RBBP8NL CABLES2 LAMA5 |
nsv953302 | Chr.20:62345345 - 62831248 on Build GRCh38 | Deletion | LOC107987285 LAMA5-AS1 LINC00659 OGFR-AS1 CABLES2 SLCO4A1-AS1 LAMA5 MIR1-1HG LOC107985428 SLCO4A1 C20orf166-AS1 OGFR NTSR1 RPS21 RBBP8NL MIR133A2 COL9A3 GATA5 MIR1-1 MRGBP |
nsv518296 | Chr.20:62203245 - 62415019 on Build GRCh38 | Loss | LOC105369209 LAMA5-AS1 ADRM1 OSBPL2 RPS21 RBBP8NL CABLES2 LAMA5 MTG2 MIR4758 HRH3 |
dgv7650n54 | Chr.20:62315508 - 62394547 on Build GRCh38 | Loss | LAMA5-AS1 RPS21 CABLES2 LAMA5 MIR4758 |
nsv586498 | Chr.20:62387568 - 62415019 on Build GRCh38 | Loss | RPS21 RBBP8NL CABLES2 |
dgv4343n100 | Chr.20:62391262 - 62569333 on Build GRCh38 | Gain | RBBP8NL MIR133A2 GATA5 CABLES2 MIR1-1HG MIR1-1 C20orf166-AS1 |
nsv834024 | Chr.20:62259914 - 62455522 on Build GRCh38 | Gain | LAMA5-AS1 ADRM1 OSBPL2 RPS21 RBBP8NL CABLES2 LAMA5 MIR4758 |
nsv470556 | Chr.20:62212004 - 62577283 on Build GRCh38 | Loss | LOC105369209 LAMA5-AS1 ADRM1 CABLES2 LAMA5 MIR1-1HG C20orf166-AS1 OSBPL2 RPS21 RBBP8NL MIR133A2 GATA5 MIR1-1 MIR4758 HRH3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |