Assay Details
Target Gene Details
Entrez Gene ID: | 51 |
Gene Name: | acyl-CoA oxidase 1 |
Gene Aliases: |
ACOX, PALMCOX, SCOX |
Location: |
Chr.17:75941507-75979434 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOX1 | NM_001185039.1 | NP_001171968.1 | ||
NM_004035.6 | NP_004026.2 | |||
NM_007292.5 | NP_009223.2 | |||
XM_011524868.2 | XP_011523170.1 | |||
XM_011524869.2 | XP_011523171.1 | |||
AK095501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3641270 | Chr.17:75929772 - 75959761 on Build GRCh38 | Gain |
![]() ![]() |
esv3641271 | Chr.17:75936747 - 75955938 on Build GRCh38 | Gain |
![]() ![]() |
nsv833545 | Chr.17:75738140 - 75945931 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)