Assay Details
Target Gene Details
Entrez Gene ID: | 863 |
Gene Name: | CBFA2/RUNX1 translocation partner 3 |
Gene Aliases: |
ETO2, MTG16, MTGR2, RUNX1T3, ZMYND4 |
Location: |
Chr.16:88874855-88977198 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CBFA2T3 | NM_005187.5 | 12 | 2044 | NP_005178.4 |
NM_175931.2 | 11 | 1710 | NP_787127.1 | |
XM_005256323.4 | 11 | 2022 | XP_005256380.1 | |
AB010419.1 | 12 | 1913 | BAA29061.1 | |
AB010420.1 | 11 | 1710 | BAA29062.1 | |
AK090985.1 | 3 | 722 | ||
AK097904.1 | 2 | 860 | ||
AK314766.1 | 12 | 1899 | ||
BC047019.1 | 12 | 1894 | ||
BC062624.1 | 12 | 1914 | AAH62624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv37n68 | Chr.16:88767460 - 88944903 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1063024 | Chr.16:88707330 - 89238482 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1928 | Chr.16:88865211 - 88897277 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
nsv471115 | Chr.16:88801844 - 89197442 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv573674 | Chr.16:88738775 - 89325907 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3639555 | Chr.16:88745961 - 88899576 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv521217 | Chr.16:88805821 - 89174375 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952071 | Chr.16:88790893 - 89019992 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3639561 | Chr.16:88837996 - 88882065 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv3065n100 | Chr.16:88711587 - 88927172 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)