Assay Details
Target Gene Details
Entrez Gene ID: | 22980 |
Gene Name: | transcription factor 25 |
Gene Aliases: |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
Location: |
Chr.16:89873583-89911384 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573744 | Chr.16:89738447 - 89915950 on Build GRCh38 | Loss | FANCA ZNF276 TCF25 LOC107984817 LOC105371419 SPIRE2 |
esv2758671 | Chr.16:89804324 - 89901541 on Build GRCh38 | Loss | FANCA TCF25 LOC107984817 LOC105371419 SPIRE2 |
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | ZNF276 CENPBD1 DEF8 TCF25 MC1R TUBB3 GAS8 SPATA2L CDK10 DPEP1 CHMP1A FANCA VPS9D1 AFG3L1P DBNDD1 GAS8-AS1 SPATA33 LOC107984817 LOC105371419 SPIRE2 VPS9D1-AS1 |
nsv573810 | Chr.16:89857498 - 90037251 on Build GRCh38 | Loss | AFG3L1P CENPBD1 DEF8 TCF25 MC1R DBNDD1 GAS8-AS1 TUBB3 GAS8 SPIRE2 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | ZNF276 CENPBD1 DEF8 SPATA2L URAHP TUBB8P7 RPL13 CHMP1A AFG3L1P DBNDD1 GAS8-AS1 SPATA33 PRDM7 LOC105371419 SPG7 SNORD68 TCF25 MC1R LOC105371414 TUBB3 GAS8 CDK10 LOC100287036 DPEP1 LOC101927817 FANCA VPS9D1 CPNE7 ANKRD11 LOC107984817 SPIRE2 VPS9D1-AS1 FAM157C ZNF778 |
nsv573799 | Chr.16:89852186 - 90022379 on Build GRCh38 | Gain | AFG3L1P CENPBD1 DEF8 TCF25 MC1R DBNDD1 TUBB3 GAS8 LOC105371419 SPIRE2 |
nsv1066499 | Chr.16:89891957 - 90028891 on Build GRCh38 | Loss | AFG3L1P CENPBD1 DEF8 TCF25 MC1R DBNDD1 TUBB3 GAS8 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC107987238 CENPBD1 URAHP TUBB8P7 RPL13 LOC105371409 CDH15 SPATA33 PIEZO1 LOC339059 SPG7 LOC400558 TCF25 MC1R LOC105371414 GAS8 SNAI3 APRT CDK10 LOC100287036 LOC101927817 FANCA SLC22A31 TRAPPC2L SPIRE2 ZNF276 LINC00304 DEF8 SPATA2L CTU2 CHMP1A SNAI3-AS1 AFG3L1P DBNDD1 GAS8-AS1 PRDM7 RNF166 CBFA2T3 LOC105371419 CYBA LOC100129697 SNORD68 TUBB3 MIR4722 LOC100289580 CDT1 ACSF3 DPEP1 VPS9D1 CPNE7 ANKRD11 GALNS IL17C MVD LOC107984817 LOC101927793 PABPN1L VPS9D1-AS1 FAM157C ZNF778 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |