Assay Details
Target Gene Details
Entrez Gene ID: | 8312 |
Gene Name: | axin 1 |
Gene Aliases: |
AXIN, PPP1R49 |
Location: |
Chr.16:287440-355226 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AXIN1 | NM_003502.3 | NP_003493.1 | ||
NM_181050.2 | NP_851393.1 | |||
NR_134879.1 | ||||
XM_011522682.2 | XP_011520984.1 | |||
XM_011522683.2 | XP_011520985.1 | |||
XM_011522684.2 | XP_011520986.1 | |||
XM_011522686.1 | XP_011520988.1 | |||
XM_017023743.1 | XP_016879232.1 | |||
XM_017023744.1 | XP_016879233.1 | |||
XM_017023745.1 | XP_016879234.1 | |||
XM_017023746.1 | XP_016879235.1 | |||
AB208876.1 | 7 | 5406 | BAD92113.1 | |
AF009674.1 | AAC51624.1 | |||
BC017447.1 | AAH17447.1 | |||
BC044648.1 | AAH44648.1 | |||
BX648449.1 | 1 | 854 | ||
KF615847.1 | ||||
KF615848.1 | ||||
KF615849.1 | ||||
KF615850.1 | ||||
KF615851.1 | ||||
KF615852.1 | ||||
KF615853.1 | ||||
KF615854.1 | ||||
KU178413.1 | ||||
KU178414.1 | ||||
KU178415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3637564 | Chr.16:287941 - 370218 on Build GRCh38 | Gain |
![]() ![]() |
nsv952896 | Chr.16:273501 - 321500 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv570951 | Chr.16:279671 - 322597 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv827477 | Chr.16:262247 - 297639 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv570947 | Chr.16:272935 - 297326 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)